missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetra-n-dodecylammonium iodide, 98%, Thermo Scientific Chemicals
$94.67 - $322.23
Chemical Identifiers
CAS | 40797-39-5 |
---|---|
Molecular Formula | C48H100IN |
Molecular Weight (g/mol) | 818.24 |
MDL Number | MFCD00051856 |
InChI Key | OBMLOTDECOTAGU-UHFFFAOYSA-M |
Synonym | tetradodecylazanium iodide, tetra-n-dodecylammonium iodide, tetradodecylammonium iodide, tetradodecylazanium,iodide |
PubChem CID | 3084801 |
SMILES | [I-].CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0815304
|
Thermo Scientific Chemicals
L0815304 |
2 g |
Each for $94.67
|
|
|||||
AAL0815309
|
Thermo Scientific Chemicals
L0815309 |
10 g |
Each for $322.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
40797-39-5 | |
818.24 | |
OBMLOTDECOTAGU-UHFFFAOYSA-M | |
3084801 |
C48H100IN | |
MFCD00051856 | |
tetradodecylazanium iodide, tetra-n-dodecylammonium iodide, tetradodecylammonium iodide, tetradodecylazanium,iodide | |
[I-].CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC |
Specifications
40797-39-5 | |
C48H100IN | |
MFCD00051856 | |
5844188 | |
tetradodecylazanium iodide, tetra-n-dodecylammonium iodide, tetradodecylammonium iodide, tetradodecylazanium,iodide | |
[I-].CCCCCCCCCCCC[N+](CCCCCCCCCCCC)(CCCCCCCCCCCC)CCCCCCCCCCCC | |
818.24 | |
818.23 | |
Tetra-n-dodecylammonium iodide |
114°C to 118°C | |
[CH3(CH2)11]4NI | |
2 g | |
Light sensitive | |
OBMLOTDECOTAGU-UHFFFAOYSA-M | |
tetradodecylazanium iodide | |
3084801 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 255-086-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only