Learn More
Tetra-n-propylammonium bromide, 98%
CAS: 1941-30-6 | C12H28BrN | 266.267 g/mol
$109.62 - $1738.82
Chemical Identifiers
| CAS | 1941-30-6 |
|---|---|
| Molecular Formula | C12H28BrN |
| Molecular Weight (g/mol) | 266.267 |
| MDL Number | MFCD00011840 |
| InChI Key | BGQMOFGZRJUORO-UHFFFAOYSA-M |
| Synonym | tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide |
| PubChem CID | 74745 |
| ChEBI | CHEBI:55318 |
| IUPAC Name | tetrapropylazanium;bromide |
| SMILES | CCC[N+](CCC)(CCC)CCC.[Br-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1152222
|
Thermo Scientific Chemicals
A1152222 |
100 g |
Each for $109.62
|
|
|||||
|
AAA1152236
|
Thermo Scientific Chemicals
A1152236 |
500 g |
Each for $365.42
|
|
|||||
|
AAA115220E
|
Thermo Scientific Chemicals
A115220E |
2500 g |
Each for $1,738.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
ApplicationsTetra-n-propylammonium bromide, is used as surface-active agents, Solvents, Intermediates, Active Ingredient for Conditioners, Antistatic Agent, Detergent Sanitisers, Softner for textiles and paper products, Phase Transfer Catalyst, Antimicrobials, Disinfection Agents And Sanitizers, Slimicidal Agents, Emulsifying Agents and Pigment Dispersers.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatabile to Strong oxidizing agents. Stable under ordinary conditions (Hygroscopic)
Chemical Identifiers
| 1941-30-6 | |
| 266.267 | |
| BGQMOFGZRJUORO-UHFFFAOYSA-M | |
| 74745 | |
| tetrapropylazanium;bromide |
| C12H28BrN | |
| MFCD00011840 | |
| tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide | |
| CHEBI:55318 | |
| CCC[N+](CCC)(CCC)CCC.[Br-] |
Specifications
| 1941-30-6 | |
| C12H28BrN | |
| 100 g | |
| Hygroscopic | |
| It is soluble in water. | |
| CCC[N+](CCC)(CCC)CCC.[Br-] | |
| 266.267 | |
| CHEBI:55318 | |
| ≥98% |
| ∼270°C (decomposition) | |
| MFCD00011840 | |
| 3567846 | |
| tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide | |
| BGQMOFGZRJUORO-UHFFFAOYSA-M | |
| tetrapropylazanium;bromide | |
| 74745 | |
| 266.27 | |
| Tetra-n-propylammonium bromide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 217-727-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only