missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabenzyl pyrophosphate, 98%, Thermo Scientific Chemicals

$396.27 - $1462.77

Chemical Identifiers

CAS 990-91-0
Molecular Formula C28H28O7P2
Molecular Weight (g/mol) 538.473
MDL Number MFCD00051941
InChI Key NSBNXCZCLRBQTA-UHFFFAOYSA-N
Synonym tetrabenzyl pyrophosphate, tetrabenzyl diphosphate, pyrophosphoric acid tetrabenzyl ester, tetrabenzylpyrophosphate, diphosphoric acid, tetrakis phenylmethyl ester, dibenzyl dibenzyloxyphosphoryl oxyphosphonate, benzyl pyrophosphate, acmc-209sbp, tetra benzyl pyrophosphate, tetrabenzyl diphosphate #
PubChem CID 563183
IUPAC Name dibenzyl bis(phenylmethoxy)phosphoryl phosphate
SMILES C1=CC=C(C=C1)COP(=O)(OCC2=CC=CC=C2)OP(=O)(OCC3=CC=CC=C3)OCC4=CC=CC=C4
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0908303
View Documents
Thermo Scientific Chemicals
L0908303
1 g
Each for $396.27
 
AAL0908306
View Documents
Thermo Scientific Chemicals
L0908306
5 g
Each for $1,462.77
 
Description

Description

Tetrabenzyl pyrophosphate is used in the preparation of phosphoryl derivatives of shikimic acid in the presence of LDA. It is used in the preparation of dibenzyl phosphoro fluoridate in the presence of cesium fluoride as a catalyst. It is also used as a precursor in pharmaceuticals and involved in the phosphorylation of inositol derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrabenzyl pyrophosphate is used in the preparation of phosphoryl derivatives of shikimic acid in the presence of LDA. It is used in the preparation of dibenzyl phosphoro fluoridate in the presence of cesium fluoride as a catalyst. It is also used as a precursor in pharmaceuticals and involved in the phosphorylation of inositol derivatives.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents, strong acid and bases and reducing agents.
Specifications

Chemical Identifiers

990-91-0
538.473
NSBNXCZCLRBQTA-UHFFFAOYSA-N
563183
C1=CC=C(C=C1)COP(=O)(OCC2=CC=CC=C2)OP(=O)(OCC3=CC=CC=C3)OCC4=CC=CC=C4
C28H28O7P2
MFCD00051941
tetrabenzyl pyrophosphate, tetrabenzyl diphosphate, pyrophosphoric acid tetrabenzyl ester, tetrabenzylpyrophosphate, diphosphoric acid, tetrakis phenylmethyl ester, dibenzyl dibenzyloxyphosphoryl oxyphosphonate, benzyl pyrophosphate, acmc-209sbp, tetra benzyl pyrophosphate, tetrabenzyl diphosphate #
dibenzyl bis(phenylmethoxy)phosphoryl phosphate

Specifications

990-91-0
C28H28O7P2
1 g
2068292
tetrabenzyl pyrophosphate, tetrabenzyl diphosphate, pyrophosphoric acid tetrabenzyl ester, tetrabenzylpyrophosphate, diphosphoric acid, tetrakis phenylmethyl ester, dibenzyl dibenzyloxyphosphoryl oxyphosphonate, benzyl pyrophosphate, acmc-209sbp, tetra benzyl pyrophosphate, tetrabenzyl diphosphate #
NSBNXCZCLRBQTA-UHFFFAOYSA-N
dibenzyl bis(phenylmethoxy)phosphoryl phosphate
563183
98%
58°C to 64°C
MFCD00051941
UN3261
Moisture sensitive
Slightly soluble in water.
C1=CC=C(C=C1)COP(=O)(OCC2=CC=CC=C2)OP(=O)(OCC3=CC=CC=C3)OCC4=CC=CC=C4
538.473
538.48
Tetrabenzyl pyrophosphate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

TSCA : No

Recommended Storage : Store at -20°C; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only