missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrabutylammonium Acetate 90.0+%, TCI America™
Supplier: TCI America T2694100G
Specifications
Tetrabutylammonium Acetate | |
98°C | |
C18H39NO2 | |
100 g | |
MCZDHTKJGDCTAE-UHFFFAOYSA-M | |
tetrabutylazanium acetate | |
82707 | |
≥90.0% (T) |
10534-59-5 | |
White-Yellow | |
MFCD00043208 | |
tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate | |
CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC | |
301.52 | |
301.52 | |
Paste |
Chemical Identifiers
10534-59-5 | |
301.52 | |
MCZDHTKJGDCTAE-UHFFFAOYSA-M | |
82707 | |
CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC |
C18H39NO2 | |
MFCD00043208 | |
tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate | |
tetrabutylazanium acetate |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : Yes