missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium Acetate 90.0+%, TCI America™

Supplier:  TCI America T269425G

 View more versions of this product

Catalog No. T269425G


Explore available promotions

Description

Description

Specifications

Specifications

Tetrabutylammonium Acetate
98°C
C18H39NO2
25 g
MCZDHTKJGDCTAE-UHFFFAOYSA-M
tetrabutylazanium acetate
82707
≥90.0% (T)
10534-59-5
White-Yellow
MFCD00043208
tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate
CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
301.52
301.52
Paste

Chemical Identifiers

10534-59-5
301.52
MCZDHTKJGDCTAE-UHFFFAOYSA-M
82707
CC([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
C18H39NO2
MFCD00043208
tetrabutylammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate, tetra-n-butylammonium acetate, tetrabutylazanium acetate, tetrabutyl ammonium acetate, 1-butanaminium, n,n,n-tributyl-, acetate 1:1, tetrabutylammonium ion acetate, 2-phosphonooxy acrylic acid cyclohexylamine salt 1:1, n,n,n-tributylbutan-1-aminiumacetate, tetrabutylazanium ethanoate
tetrabutylazanium acetate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions