missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Chloride 98.0+%, TCI America™
$70.84 - $553.62
Chemical Identifiers
| CAS | 1112-67-0 |
|---|---|
| Molecular Formula | C16H36ClN |
| Molecular Weight (g/mol) | 277.921 |
| MDL Number | MFCD00011635 |
| InChI Key | NHGXDBSUJJNIRV-UHFFFAOYSA-M |
| Synonym | tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride |
| PubChem CID | 70681 |
| ChEBI | CHEBI:51988 |
| IUPAC Name | tetrabutylazanium;chloride |
| SMILES | CCCC[N+](CCCC)(CCCC)CCCC.[Cl-] |
Chemical Identifiers
| 1112-67-0 | |
| 277.921 | |
| NHGXDBSUJJNIRV-UHFFFAOYSA-M | |
| 70681 | |
| tetrabutylazanium;chloride |
| C16H36ClN | |
| MFCD00011635 | |
| tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride | |
| CHEBI:51988 | |
| CCCC[N+](CCCC)(CCCC)CCCC.[Cl-] |
Specifications
| 1112-67-0 | |
| White | |
| MFCD00011635 | |
| tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride | |
| CCCC[N+](CCCC)(CCCC)CCCC.[Cl-] | |
| 277.921 | |
| CHEBI:51988 | |
| ≥98.0% (T) | |
| Tetrabutylammonium Chloride |
| 70°C | |
| C16H36ClN | |
| 5 g | |
| NHGXDBSUJJNIRV-UHFFFAOYSA-M | |
| tetrabutylazanium;chloride | |
| 70681 | |
| 277.92 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : Yes