missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium hydrogen sulfate, 98%, Thermo Scientific Chemicals

$104.90 - $922.28

Chemical Identifiers

CAS 32503-27-8
Molecular Formula C16H37NO4S
Molecular Weight (g/mol) 339.54
MDL Number MFCD00011637
InChI Key SHFJWMWCIHQNCP-UHFFFAOYSA-M
Synonym tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
PubChem CID 94433
IUPAC Name hydrogen sulfate;tetrabutylazanium
SMILES OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC168380250
SDS View Documents
Thermo Scientific Chemicals
168380250
25 g Plastic bottle
Each for $104.90
 
AC168381000
SDS View Documents
Thermo Scientific Chemicals
168381000
100 g Plastic bottle
Each for $281.10
 
AC168382500
SDS View Documents
Thermo Scientific Chemicals
168382500
250 g Plastic bottle
Each for $651.32
 
AC168380010
SDS View Documents
Thermo Scientific Chemicals
168380010
1 kg Plastic bottle
Each for $922.28
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Chemical Identifiers

32503-27-8
339.54
SHFJWMWCIHQNCP-UHFFFAOYSA-M
94433
OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
C16H37NO4S
MFCD00011637
tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
hydrogen sulfate;tetrabutylazanium

Specifications

32503-27-8
White to Beige
98%
C16H37NO4S
MFCD00011637
06,565; 07,354; 13,94
Solubility in water: >50%.
OS([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC
339.54
339.53
Crystalline Powder and/or Chunks
168.0°C to 172.0°C
Authentic
Plastic bottle
[CH3(CH2)3]4NHSO4
25 g
tetrabutylammonium hydrogen sulfate, tetrabutylammonium hydrogensulfate, tetrabutylammonium bisulfate, tetra-n-butylammonium hydrogen sulfate, tbahs, tetrabutylammonium sulfate 1:1, 1-butanaminium, n,n,n-tributyl-, sulfate 1:1, tetrabutylammonium hydrogen sulphate, tetrabutylazanium hydrogen sulfate, ipc-tba-hs
SHFJWMWCIHQNCP-UHFFFAOYSA-M
hydrogen sulfate;tetrabutylazanium
94433
98%
Tetrabutylammonium hydrogen sulfate, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 251-068-5

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only