missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrabutylammonium Hydroxide, 1.0 M Solution in Methanol, Spectrum™ Chemical
$537.59 - $2895.09
Chemical Identifiers
CAS | 2052-49-5 |
---|---|
Molecular Formula | C16H37NO |
Molecular Weight (g/mol) | 259.48 |
MDL Number | MFCD00009425 |
InChI Key | VDZOOKBUILJEDG-UHFFFAOYSA-M |
IUPAC Name | tetrabutylazanium hydroxide |
SMILES | [OH-].CCCC[N+](CCCC)(CCCC)CCCC |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
18614342
|
Spectrum Chemical Mfg Cor
T105825ML |
25 mL | Amber Glass Bottle |
Each for $537.59
|
|
||||
18614343
|
Spectrum Chemical Mfg Cor
T1058100ML |
100 mL | Amber Glass Bottle |
Each for $1,146.32
|
|
||||
18614344
|
Spectrum Chemical Mfg Cor
T1058500MLGL |
500 mL | Amber Glass Bottle |
Each for $2,895.09
|
|
||||
Chemical Identifiers
2052-49-5 | |
259.48 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC |
C16H37NO | |
MFCD00009425 | |
tetrabutylazanium hydroxide |
Specifications
2052-49-5,7732-18-5 | |
Amber Glass Bottle | |
MFCD00009425 | |
VDZOOKBUILJEDG-UHFFFAOYSA-M | |
tetrabutylazanium hydroxide | |
Reagent |
1 | |
C16H37NO | |
25 mL | |
[OH-].CCCC[N+](CCCC)(CCCC)CCCC | |
259.48 | |
0.9-1.1 M |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.