missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium p-Nitrophenoxide 98.0+%, TCI America™

Supplier:  TCI America T266925G

 View more versions of this product

Catalog No. T266925G


Explore available promotions

Description

Description

Specifications

Specifications

Tetrabutylammonium p-Nitrophenoxide
145°C
C22H40N2O3
JWRHOHUAHHYNBL-UHFFFAOYSA-M
4-nitrophenolate;tetrabutylazanium
18148
≥98.0% (HPLC,T)
3002-48-0
Yellow
25 g
CCCC[N+](CCCC)(CCCC)CCCC.C1=CC(=CC=C1[N+](=O)[O-])[O-]
380.573
380.57
Crystalline Powder

Chemical Identifiers

3002-48-0
380.573
18148
CCCC[N+](CCCC)(CCCC)CCCC.C1=CC(=CC=C1[N+](=O)[O-])[O-]
C22H40N2O3
JWRHOHUAHHYNBL-UHFFFAOYSA-M
4-nitrophenolate;tetrabutylazanium
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions