missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrabutylammonium phosphate monobasic, 0.5M conc. in HPLC grade water, HPLC grade, Thermo Scientific Chemicals
$68.36 - $621.03
Specifications
Boiling Point | 100.0°C |
---|---|
Chemical Name or Material | Tetrabutylammonium phosphate monobasic |
CAS | 5574-97-0,7732-18-5 |
Physical Form | Solution |
Molecular Formula | C16H38NO4P |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC420170250
|
Thermo Scientific Chemicals
420170250 |
25 mL | Glass bottle |
Each for $68.36
|
|
||||
AC420171000
|
Thermo Scientific Chemicals
420171000 |
100 mL | Glass bottle |
Each for $156.23
|
|
||||
AC420175000
|
Thermo Scientific Chemicals
420175000 |
500 mL | Glass bottle |
Each for $621.03
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography, Phase transfer catalystSpecifications
100.0°C | |
5574-97-0,7732-18-5 | |
C16H38NO4P | |
MFCD00064526 | |
ARRNBPCNZJXHRJ-UHFFFAOYSA-M | |
dihydrogen phosphate;tetrabutylazanium | |
2735142 | |
0.48 to 0.52M | |
1.01 |
Tetrabutylammonium phosphate monobasic | |
Solution | |
(CH3(CH2)3)4NH2PO4 | |
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion | |
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC | |
339.46 | |
339.45 | |
Glass bottle | |
1.0100g/mL |
RUO – Research Use Only