missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrabutylammonium Phosphate (White to Off-White Powder/HPLC Grade), Fisher Chemical™

Improve HPLC separation of acidic analytes by ion pairing.

Supplier:  Thermo Fisher Scientific O6117100

 View more versions of this product

Catalog No. O6117100



Explore available promotions

Description

Description

  • High purity
  • Application tested
  • Dissolves easily
Specifications

Specifications

Tetrabutylammonium Phosphate
0.02 AU max. at 214nm, 0.1 AU max. at 210nm, 0.2 AU max. at 200nm
White
≥98.5 %
(CH3CH2CH2CH2)4NH2PO4
100 g
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
339.46
339.45
HPLC
5574-97-0
148°C
4 to 5
C16H38NO4P
MFCD00064526
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
tetrabutylazanium dihydrogen phosphate
2735142
≥98.5%
Powder or Solid

Chemical Identifiers

5574-97-0
339.46
ARRNBPCNZJXHRJ-UHFFFAOYSA-M
tetrabutylazanium dihydrogen phosphate
C16H38NO4P
MFCD00064526
2735142
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC
Videos
Safety and Handling

Safety and Handling

SDS Documents

Documents

Promotions

Promotions