missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrabutylammonium Tetrafluoroborate 98.0+%, TCI America™
Supplier: TCI America T0914500G
Specifications
Tetrabutylammonium Tetrafluoroborate | |
161°C | |
C16H36BF4N | |
500 g | |
tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4; | |
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC | |
329.27 | |
329.27 | |
Crystalline Powder |
429-42-5 | |
White | |
MFCD00011634 | |
1759 | |
NNZZSJSQYOFZAM-UHFFFAOYSA-N | |
tetrabutylazanium; tetrafluoroboranuide | |
67932 | |
≥98.0% (N) |
Chemical Identifiers
429-42-5 | |
329.27 | |
NNZZSJSQYOFZAM-UHFFFAOYSA-N | |
67932 | |
F[B-](F)(F)F.CCCC[N+](CCCC)(CCCC)CCCC |
C16H36BF4N | |
MFCD00011634 | |
tetrabutylammonium tetrafluoroborate, tetra-n-butylammonium tetrafluoroborate, tetrabutylammonium fluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-, tetrabutyl ammonium tetrafluoroborate, tetrabutylammonium ion tetrafluoroborate, 1-butanaminium, n,n,n-tributyl-, tetrafluoroborate 1-1:1, tbabf4, acmc-209jry, tbabf4; | |
tetrabutylazanium; tetrafluoroboranuide |
Safety and Handling
EINECSNumber : (2)-0186
TSCA : Yes