missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gold Chloride Trihydrate, Certified ACS, Fisher Chemical™
Supplier: Thermo Fisher Scientific G545
Description
- Used as a gold catalyst or source of gold for organic reactions
- Intended for research use only. Not for human or animal diagnostic purposes.
Specifications
Tetrachloroauric Acid Trihydrate | |
Yellow | |
Solid | |
AuCl4 | |
Gold(III) chloride trihydrate, Tetrachloroauric acid trihydrate, Hydrogren tetrachloroaurate(III), Tetrachloroauric(III) acid, Chloroauric acid trihydrate | |
Cl[Au](Cl)(Cl)Cl | |
338.77 | |
≥49% | |
Pass Test | |
3.9g/cm³ |
254°C | |
16961-25-4 | |
5 g | |
MFCD00149904 | |
CQVDKGFMVXRRAI-UHFFFAOYSA-J | |
tetrachlorogold | |
393.83 | |
Certified ACS | |
Amber Glass |
Chemical Identifiers
16961-25-4 | |
338.77 | |
CQVDKGFMVXRRAI-UHFFFAOYSA-J | |
tetrachlorogold |
AuCl4 | |
MFCD00149904 | |
Gold(III) chloride trihydrate, Tetrachloroauric acid trihydrate, Hydrogren tetrachloroaurate(III), Tetrachloroauric(III) acid, Chloroauric acid trihydrate | |
Cl[Au](Cl)(Cl)Cl |
Safety and Handling
Recommended Storage : Refrigerate