missing translation for 'onlineSavingsMsg'
Learn More

Tetrachlorobis(tetrahydrofuran)zirconium(IV), Thermo Scientific Chemicals

$254.21 - $254.21

Chemical Identifiers

CAS 21959-01-3
Molecular Formula C8H16Cl4O2Zr
Molecular Weight (g/mol) 377.238
MDL Number MFCD00145367
InChI Key VDJJKYYENYIHFF-UHFFFAOYSA-J
Synonym tetrachlorobis tetrahydrofuran zirconium, zrcl4 thf 2, oxolane; tetrachlorozirconium, tetrachlorobis tetrahydrofurane zirconium, zirconium tetrachloride tetrahydrofuran complex, zirconium iv chloride tetrahydrofuran complex, bis tetrahydrofuran ; zirconium iv chloride, zirconium iv chloride tetrahydrofuran complex 1:2
PubChem CID 2734040
IUPAC Name oxolane;tetrachlorozirconium
SMILES C1CCOC1.C1CCOC1.Cl[Zr](Cl)(Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
AA3955009
View Documents
Thermo Scientific Chemicals
03955009
10 g
Each for $254.21
Only null left
 
Description

Description

It is an important organic intermediate and can be used in agrochemicals, paharmaceuticals and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

21959-01-3
377.238
VDJJKYYENYIHFF-UHFFFAOYSA-J
2734040
C1CCOC1.C1CCOC1.Cl[Zr](Cl)(Cl)Cl
C8H16Cl4O2Zr
MFCD00145367
tetrachlorobis tetrahydrofuran zirconium, zrcl4 thf 2, oxolane; tetrachlorozirconium, tetrachlorobis tetrahydrofurane zirconium, zirconium tetrachloride tetrahydrofuran complex, zirconium iv chloride tetrahydrofuran complex, bis tetrahydrofuran ; zirconium iv chloride, zirconium iv chloride tetrahydrofuran complex 1:2
oxolane;tetrachlorozirconium

Specifications

175°C to 177°C
White
Crystalline
MFCD00145367
tetrachlorobis tetrahydrofuran zirconium, zrcl4 thf 2, oxolane; tetrachlorozirconium, tetrachlorobis tetrahydrofurane zirconium, zirconium tetrachloride tetrahydrofuran complex, zirconium iv chloride tetrahydrofuran complex, bis tetrahydrofuran ; zirconium iv chloride, zirconium iv chloride tetrahydrofuran complex 1:2
VDJJKYYENYIHFF-UHFFFAOYSA-J
oxolane;tetrachlorozirconium
2734040
Tetrachlorobis(tetrahydrofuran)zirconium(IV)
21959-01-3
10 g
C8H16Cl4O2Zr
UN2925
Reacts with water.
C1CCOC1.C1CCOC1.Cl[Zr](Cl)(Cl)Cl
377.238
377.25
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H314-H318
Flammable solid.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

RUO – Research Use Only