Learn More
Tetrachlorophthalic anhydride, 98%, Thermo Scientific Chemicals
$96.96 - $1015.36
Chemical Identifiers
CAS | 117-08-8 |
---|---|
Molecular Formula | C8Cl4O3 |
Molecular Weight (g/mol) | 285.885 |
MDL Number | MFCD00005920 |
InChI Key | AUHHYELHRWCWEZ-UHFFFAOYSA-N |
Synonym | tetrachlorophthalic anhydride, tetrathal, 4,5,6,7-tetrachloro-1,3-isobenzofurandione, niagathal, 1,3-isobenzofurandione, 4,5,6,7-tetrachloro, phthalic anhydride, tetrachloro, 4,5,6,7-tetrachloroisobenzofuran-1,3-dione, 1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran, unii-76glw0lbek, ccris 6202 |
PubChem CID | 8326 |
ChEBI | CHEBI:59097 |
IUPAC Name | 4,5,6,7-tetrachloro-2-benzofuran-1,3-dione |
SMILES | C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1740436
|
Thermo Scientific Chemicals
A1740436 |
500 g |
Each for $96.96
|
|
|||||
AAA174040E
|
Thermo Scientific Chemicals
A174040E |
2500 g |
Each for $326.89
|
|
|||||
AAA174040C
|
Thermo Scientific Chemicals
A174040C |
10,000 g |
Each for $1,015.36
|
|
|||||
Description
Protection of amines as their tetrachlorophthalimide derivatives, by reaction with the anhydride in the presence of triethylamine, has been found to be useful in oligosaccharide chemistry. The group is stable to a wide range of reagents but can be selectively cleaved with ethylenediamine at lower temperatures than for the parent phthalimide derivatives; in particular, carboxylic esters are not cleaved under these conditions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsProtection of amines as their tetrachlorophthalimide derivatives, by reaction with the anhydride in the presence of triethylamine, has been found to be useful in oligosaccharide chemistry. The group is stable to a wide range of reagents but can be selectively cleaved with ethylenediamine at lower temperatures than for the parent phthalimide derivatives; in particular, carboxylic esters are not cleaved under these conditions.
Solubility
Hydrolyzes slowly in water.
Notes
Hygroscopic. Store away from oxidizing agents, acids.
Chemical Identifiers
117-08-8 | |
285.885 | |
AUHHYELHRWCWEZ-UHFFFAOYSA-N | |
8326 | |
4,5,6,7-tetrachloro-2-benzofuran-1,3-dione |
C8Cl4O3 | |
MFCD00005920 | |
tetrachlorophthalic anhydride, tetrathal, 4,5,6,7-tetrachloro-1,3-isobenzofurandione, niagathal, 1,3-isobenzofurandione, 4,5,6,7-tetrachloro, phthalic anhydride, tetrachloro, 4,5,6,7-tetrachloroisobenzofuran-1,3-dione, 1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran, unii-76glw0lbek, ccris 6202 | |
CHEBI:59097 | |
C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O |
Specifications
117-08-8 | |
349°C to 354°C | |
Weak | |
MFCD00005920 | |
UN3077 | |
Moisture sensitive | |
Hydrolyzes slowly in water. | |
C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O | |
285.885 | |
CHEBI:59097 | |
98% |
254°C to 258°C | |
362°C (683°F) | |
C8Cl4O3 | |
500 g | |
211560 | |
tetrachlorophthalic anhydride, tetrathal, 4,5,6,7-tetrachloro-1,3-isobenzofurandione, niagathal, 1,3-isobenzofurandione, 4,5,6,7-tetrachloro, phthalic anhydride, tetrachloro, 4,5,6,7-tetrachloroisobenzofuran-1,3-dione, 1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran, unii-76glw0lbek, ccris 6202 | |
AUHHYELHRWCWEZ-UHFFFAOYSA-N | |
4,5,6,7-tetrachloro-2-benzofuran-1,3-dione | |
8326 | |
285.9 | |
Tetrachlorophthalic anhydride |
Safety and Handling
GHS H Statement
H334-H318-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes serious eye damage.
May cause an allergic skin reaction.
P201-P202-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P305+P351+P338-P308+P313-P310-P333+P313-P342+P311-P363-P501c
H317-H318-H334-H350
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 204-171-4
RTECSNumber : TI3450000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only