missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetracyanoethylene, 98%, Thermo Scientific Chemicals

 

$236.21 - $528.80

Chemical Identifiers

CAS 670-54-2
Molecular Formula C6N4
Molecular Weight (g/mol) 128.09
InChI Key NLDYACGHTUPAQU-UHFFFAOYSA-N
Synonym tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene
PubChem CID 12635
IUPAC Name ethene-1,1,2,2-tetracarbonitrile
SMILES C(#N)C(=C(C#N)C#N)C#N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC138050100
SDS View Documents
Thermo Scientific Chemicals
138050100
10 g Glass bottle
Each for $236.21
 
AC138050250
SDS View Documents
Thermo Scientific Chemicals
138050250
25 g Glass bottle
Each for $528.80
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
Specifications

Chemical Identifiers

670-54-2
128.09
tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene
ethene-1,1,2,2-tetracarbonitrile
C6N4
NLDYACGHTUPAQU-UHFFFAOYSA-N
12635
C(#N)C(=C(C#N)C#N)C#N

Specifications

670-54-2
223.0°C
97.5% min. (Ex N)
C6N4
10 g
15,934
Solubility in water: hydrolyzes (forming hcn) in water. Other solubilities: slightly soluble in: ether, benzene,, carbon tetrachloride, soluble in: acetone
C(#N)C(=C(C#N)C#N)C#N
128.09
128.09
Crystalline Powder, Crystals or Chunks
196.0°C to 201.0°C
Authentic
Glass bottle
(NC)2C=C(CN)2
01,1133; 02,397; 05,647; 06,567; 13,289
tetracyanoethylene, tetracyanoethene, ethylenetetracarbonitrile, tcne, ethenetetracarbonitrile, ethene, tetracyano, 1,1,2,2-ethenetetracarbonitrile, tetrakyanethylen, 1,1,2,2-tetracyanoethene, 1,1,2,2-tetracyanoethylene
NLDYACGHTUPAQU-UHFFFAOYSA-N
ethene-1,1,2,2-tetracarbonitrile
12635
98%
Tetracyanoethylene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal in contact with skin.
Fatal if swallowed.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Rinse mouth.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable

GHS Signal Word: Danger

EINECSNumber : 211-578-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only