missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetradecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt 98.0+%, TCI America™
Supplier: TCI America T265325G
Specifications
Tetradecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt [for Biochemical Research] | |
245°C | |
C19H41NO3S | |
25 g | |
BHATUINFZWUDIX-UHFFFAOYSA-N | |
3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate | |
84705 | |
363.60 | |
Crystalline Powder |
14933-09-6 | |
White | |
MFCD00036910 | |
myristyl sulfobetaine, zwittergent 3-14, n-tetradecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dimethyl tetradecyl ammonio propane-1-sulfonate, 3-n,n-dimethylmyristylammonio propanesulfonate, dimethyl 3-sulphonatopropyl tetradecylammonium, zwittergent™ 3-14 detergent, 3-myristyldimethylammonio propanesulfonate, 3-n,n-dimethyltetradecylammonio propanesulfonate | |
CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] | |
363.601 | |
CHEBI:10129 | |
≥98.0% (N) |
Chemical Identifiers
14933-09-6 | |
363.601 | |
BHATUINFZWUDIX-UHFFFAOYSA-N | |
84705 | |
3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate |
C19H41NO3S | |
MFCD00036910 | |
myristyl sulfobetaine, zwittergent 3-14, n-tetradecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dimethyl tetradecyl ammonio propane-1-sulfonate, 3-n,n-dimethylmyristylammonio propanesulfonate, dimethyl 3-sulphonatopropyl tetradecylammonium, zwittergent™ 3-14 detergent, 3-myristyldimethylammonio propanesulfonate, 3-n,n-dimethyltetradecylammonio propanesulfonate | |
CHEBI:10129 | |
CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-] |
Safety and Handling
TSCA : No