missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetradecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt 98.0+%, TCI America™

Supplier:  TCI America T26535G

 View more versions of this product

Catalog No. T26535G


Explore available promotions

Description

Description

Specifications

Specifications

Tetradecyldimethyl(3-sulfopropyl)ammonium Hydroxide Inner Salt [for Biochemical Research]
245°C
C19H41NO3S
5 g
BHATUINFZWUDIX-UHFFFAOYSA-N
3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate
84705
363.60
Crystalline Powder
14933-09-6
White
MFCD00036910
myristyl sulfobetaine, zwittergent 3-14, n-tetradecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dimethyl tetradecyl ammonio propane-1-sulfonate, 3-n,n-dimethylmyristylammonio propanesulfonate, dimethyl 3-sulphonatopropyl tetradecylammonium, zwittergent™ 3-14 detergent, 3-myristyldimethylammonio propanesulfonate, 3-n,n-dimethyltetradecylammonio propanesulfonate
CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]
363.601
CHEBI:10129
≥98.0% (N)

Chemical Identifiers

14933-09-6
363.601
BHATUINFZWUDIX-UHFFFAOYSA-N
84705
3-[dimethyl(tetradecyl)azaniumyl]propane-1-sulfonate
C19H41NO3S
MFCD00036910
myristyl sulfobetaine, zwittergent 3-14, n-tetradecyl-n,n-dimethyl-3-ammonio-1-propanesulfonate, 3-dimethyl tetradecyl ammonio propane-1-sulfonate, 3-n,n-dimethylmyristylammonio propanesulfonate, dimethyl 3-sulphonatopropyl tetradecylammonium, zwittergent™ 3-14 detergent, 3-myristyldimethylammonio propanesulfonate, 3-n,n-dimethyltetradecylammonio propanesulfonate
CHEBI:10129
CCCCCCCCCCCCCC[N+](C)(C)CCCS(=O)(=O)[O-]
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions