missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethyl (dimethylaminomethylene)diphosphonate, 97%, Thermo Scientific Chemicals

$98.00 - $302.18

Chemical Identifiers

CAS 18855-52-2
Molecular Formula C11H27NO6P2
Molecular Weight (g/mol) 331.286
MDL Number MFCD00015128
InChI Key SQDDGOVNJMIFFT-UHFFFAOYSA-N
Synonym tetraethyl dimethylaminomethylenediphosphonate, tetraethyl dimethylaminomethylene diphosphonate, phosphonic acid, dimethylamino methylene bis-, tetraethyl ester, tetraethyl dimethylamino methylene bisphosphonate, acmc-1bs7f, 1,1-bis diethoxyphosphoryl-n,n-dimethylmethanamine, dimethylamino methylene bis phosphonic acid diethyl ester, diethyl diethoxyphosphoryl dimethylamino methylphosphonate, diethoxy-phosphoryl-dimethylamino-methyl-phosphonic acid diethyl ester
PubChem CID 86795
IUPAC Name 1,1-bis(diethoxyphosphoryl)-N,N-dimethylmethanamine
SMILES CCOP(=O)(C(N(C)C)P(=O)(OCC)OCC)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1458106
View Documents
Thermo Scientific Chemicals
A1458106
5 g
Each for $98.00
 
AAA1458114
View Documents
Thermo Scientific Chemicals
A1458114
25 g
Each for $302.18
 
Description

Description

Tetraethyl (dimethylaminomethylene)diphosphonate undergoes Horner-Wadsworth-Emmons olefination with an aldehyde gives the ?-phosphonato enamine, readily convertible to the homologous acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethyl (dimethylaminomethylene)diphosphonate undergoes Horner-Wadsworth-Emmons olefination with an aldehyde gives the ɑ-phosphonato enamine, readily convertible to the homologous acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

18855-52-2
331.286
SQDDGOVNJMIFFT-UHFFFAOYSA-N
86795
CCOP(=O)(C(N(C)C)P(=O)(OCC)OCC)OCC
C11H27NO6P2
MFCD00015128
tetraethyl dimethylaminomethylenediphosphonate, tetraethyl dimethylaminomethylene diphosphonate, phosphonic acid, dimethylamino methylene bis-, tetraethyl ester, tetraethyl dimethylamino methylene bisphosphonate, acmc-1bs7f, 1,1-bis diethoxyphosphoryl-n,n-dimethylmethanamine, dimethylamino methylene bis phosphonic acid diethyl ester, diethyl diethoxyphosphoryl dimethylamino methylphosphonate, diethoxy-phosphoryl-dimethylamino-methyl-phosphonic acid diethyl ester
1,1-bis(diethoxyphosphoryl)-N,N-dimethylmethanamine

Specifications

18855-52-2
C11H27NO6P2
MFCD00015128
1914890
Not miscible or difficult to mix in water.
CCOP(=O)(C(N(C)C)P(=O)(OCC)OCC)OCC
331.286
331.29
Tetraethyl (dimethylaminomethylene)diphosphonate
138°C to 140°C (1 mmHg)
1.451
5 g
tetraethyl dimethylaminomethylenediphosphonate, tetraethyl dimethylaminomethylene diphosphonate, phosphonic acid, dimethylamino methylene bis-, tetraethyl ester, tetraethyl dimethylamino methylene bisphosphonate, acmc-1bs7f, 1,1-bis diethoxyphosphoryl-n,n-dimethylmethanamine, dimethylamino methylene bis phosphonic acid diethyl ester, diethyl diethoxyphosphoryl dimethylamino methylphosphonate, diethoxy-phosphoryl-dimethylamino-methyl-phosphonic acid diethyl ester
SQDDGOVNJMIFFT-UHFFFAOYSA-N
1,1-bis(diethoxyphosphoryl)-N,N-dimethylmethanamine
86795
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 242-627-4

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only