missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethyl methylenediphosphonate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1453214

 View more versions of this product

Catalog No. AAA1453214


Explore available promotions

Description

Description

Tetraethyl methylidenediphosphonate is used in the synthesis of tetrazoloquinoline-based mono- and bisphosphonate esters. It acts as an intermediate for the synthesis of bisphosphonate and bisphosphonic acid derivatives with anticancer and antischistosomal activity. It is also used as a reactant for synthesis of dual substrate-site inhibitors of 3-deoxy-D-arabino-heputosonate 7-phosphate synthase and alpha-amino acid derivatives containing gem-biophosphonates, Lycopene through Wittig-Horner reaction. It is also used as a precursor for the preparation of dendritic polyglycerol anions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethyl methylidenediphosphonate is used in the synthesis of tetrazoloquinoline-based mono- and bisphosphonate esters. It acts as an intermediate for the synthesis of bisphosphonate and bisphosphonic acid derivatives with anticancer and antischistosomal activity. It is also used as a reactant for synthesis of dual substrate-site inhibitors of 3-deoxy-D-arabino-heputosonate 7-phosphate synthase and alpha-amino acid derivatives containing gem-biophosphonates, Lycopene through Wittig-Horner reaction. It is also used as a precursor for the preparation of dendritic polyglycerol anions.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Tetraethyl methylenediphosphonate
1.162
>110°C (230°F)
1.44
25 g
tetraethyl methylenediphosphonate, tetraethyl methanediphosphonate, tetraethyl diphosphonomethane, methylenebis diethylphosphonate, tetraethyl methylenebisphosphonate, phosphonic acid, methylenebis-, tetraethyl ester, methylenebis diethoxyphosphine oxide, methylenebisphosphonic acid, tetraethyl ester, phosphonic acid, methylenedi-, tetraethyl ester, methylenedi phosphonic acid tetraethyl ester
STJWVOQLJPNAQL-UHFFFAOYSA-N
1-[diethoxyphosphorylmethyl(ethoxy)phosphoryl]oxyethane
15455
≥98%
1660-94-2
151°C to 153°C (1.2 mmHg)
C9H22O6P2
MFCD00039887
1813241
Slightly miscible with water.
CCOP(=O)(CP(=O)(OCC)OCC)OCC
288.22
288.22

Chemical Identifiers

1660-94-2
288.22
STJWVOQLJPNAQL-UHFFFAOYSA-N
15455
C9H22O6P2
MFCD00039887
tetraethyl methylenediphosphonate, tetraethyl methanediphosphonate, tetraethyl diphosphonomethane, methylenebis diethylphosphonate, tetraethyl methylenebisphosphonate, phosphonic acid, methylenebis-, tetraethyl ester, methylenebis diethoxyphosphine oxide, methylenebisphosphonic acid, tetraethyl ester, phosphonic acid, methylenedi-, tetraethyl ester, methylenedi phosphonic acid tetraethyl ester
CCOP(=O)(CP(=O)(OCC)OCC)OCC
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 216-764-5

RTECSNumber : SZ9150000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only