missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyl p-Xylylenediphosphonate 95.0+%, TCI America™

Supplier:  TCI America T1582100G

 View more versions of this product

Catalog No. T1582100G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Tetraethyl p-Xylylenediphosphonate
76°C
220°C
MFCD00671576
p-Bis(diethylphosphono)xylene, p-Xylylenediphosphonic Acid Tetraethyl Ester
CCOP(=O)(CC1=CC=C(C=C1)CP(=O)(OCC)OCC)OCC
378.34
378.34
Crystalline Powder
4546-04-7
White
C16H28O6P2
100 g
XTKQUBKFKSHRPS-UHFFFAOYSA-N
1,4-bis(diethoxyphosphorylmethyl)benzene
78303
≥95.0% (HPLC)

Chemical Identifiers

4546-04-7
378.34
XTKQUBKFKSHRPS-UHFFFAOYSA-N
78303
CCOP(=O)(CC1=CC=C(C=C1)CP(=O)(OCC)OCC)OCC
C16H28O6P2
MFCD00671576
p-Bis(diethylphosphono)xylene, p-Xylylenediphosphonic Acid Tetraethyl Ester
1,4-bis(diethoxyphosphorylmethyl)benzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions