Learn More
Tetraethylammonium chloride hydrate, 98%, Thermo Scientific Chemicals
$59.40 - $570.15
Chemical Identifiers
CAS | 68696-18-4 |
---|---|
Molecular Formula | C8H22ClNO |
Molecular Weight (g/mol) | 183.72 |
MDL Number | MFCD00149992 |
InChI Key | CALLTGJPWMIDPC-UHFFFAOYSA-M |
Synonym | tetraethylammonium chloride hydrate, tetraethylammonium chloride monohydrate, tetraethylazanium chloride hydrate, tetraethylammonium hydrate chloride, c8h20n.cl.h2o, tetraethylammoniumchloride hydrate, tetraethyl ammonium chloride hydrate, tetraethylammonium chloride hydrate, cryst, n,n,n-triethylethanaminium chloride-water 1/1/1, tetraethylammonium chloride, monohydrate |
PubChem CID | 16211205 |
IUPAC Name | tetraethylazanium;chloride;hydrate |
SMILES | CC[N+](CC)(CC)CC.O.[Cl-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1521514
|
Thermo Scientific Chemicals
A1521514 |
25 g |
Each for $59.40
|
|
|||||
AAA1521522
|
Thermo Scientific Chemicals
A1521522 |
100 g |
Each for $143.14
|
|
|||||
AAA1521536
|
Thermo Scientific Chemicals
A1521536 |
500 g |
Each for $570.15
|
|
|||||
Description
Tetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetraethylammonium chloride monohydrate is used as pharmaceutical intermediate.
Solubility
Soluble in water (1410 g/L at 20°C).
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
68696-18-4 | |
183.72 | |
CALLTGJPWMIDPC-UHFFFAOYSA-M | |
16211205 | |
CC[N+](CC)(CC)CC.O.[Cl-] |
C8H22ClNO | |
MFCD00149992 | |
tetraethylammonium chloride hydrate, tetraethylammonium chloride monohydrate, tetraethylazanium chloride hydrate, tetraethylammonium hydrate chloride, c8h20n.cl.h2o, tetraethylammoniumchloride hydrate, tetraethyl ammonium chloride hydrate, tetraethylammonium chloride hydrate, cryst, n,n,n-triethylethanaminium chloride-water 1/1/1, tetraethylammonium chloride, monohydrate | |
tetraethylazanium;chloride;hydrate |
Specifications
68696-18-4 | |
1.0801 | |
MFCD00149992 | |
3563247 | |
tetraethylammonium chloride hydrate, tetraethylammonium chloride monohydrate, tetraethylazanium chloride hydrate, tetraethylammonium hydrate chloride, c8h20n.cl.h2o, tetraethylammoniumchloride hydrate, tetraethyl ammonium chloride hydrate, tetraethylammonium chloride hydrate, cryst, n,n,n-triethylethanaminium chloride-water 1/1/1, tetraethylammonium chloride, monohydrate | |
CALLTGJPWMIDPC-UHFFFAOYSA-M | |
tetraethylazanium;chloride;hydrate | |
16211205 | |
98% |
∼110°C | |
C8H22ClNO | |
25 g | |
14,9200 | |
Soluble in water (1410g/L at 20°C). | |
CC[N+](CC)(CC)CC.O.[Cl-] | |
183.72 | |
183.72 (165.71 Anhydrous) | |
Tetraethylammonium chloride monohydrate |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 200-267-5
RTECSNumber : BS6125000
TSCA : Yes
RUO – Research Use Only