missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium hydroxide, 20 wt.% in water

CAS: 77-98-5 | C8H21NO | 147.26 g/mol

Supplier:  Thermo Scientific Chemicals 176072500

 View more versions of this product

Catalog No. AC176072500


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetraethylammonium hydroxide
77-98-5,7732-18-5
1.0200g/mL
Plastic bottle
1.402
MFCD00009024
1.02
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
LRGJRHZIDJQFCL-UHFFFAOYSA-M
tetraethylazanium;hydroxide
6509
≥20%
20 wt.% in Water
Colorless to Yellow
102.0°C
C8H21NO
(C2H5)4NOH
250 mL
15, 9346
Solubility in water: soluble.
[OH-].CC[N+](CC)(CC)CC
147.26
147.26
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only