missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylammonium nitrate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 420310250

 View more versions of this product

Catalog No. AC420310250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetraethylammonium nitrate
1941-26-0
White
99%
C8H20N2O3
MFCD00041978
tetraethylammonium nitrate, ethanaminium, n,n,n-triethyl-, nitrate, ammonium, tetraethyl-, nitrate, ethanaminium, n,n,n-triethyl-, nitrate 1:1, tetraethylazanium nitrate, tetraethyl ammonium nitrate, tetraethylammonium nitrate w/w aqueous solution
JTJKNAJRGLQKDZ-UHFFFAOYSA-N
tetraethylazanium;nitrate
74744
99%
99%
>240.0°C
Authentic
Glass bottle
(C2H5)4NNO3
25 g
Solubility in water: >50% (20°C).
[O-][N+]([O-])=O.CC[N+](CC)(CC)CC
192.26
192.26
Crystalline Powder

Chemical Identifiers

1941-26-0
192.26
JTJKNAJRGLQKDZ-UHFFFAOYSA-N
74744
[O-][N+]([O-])=O.CC[N+](CC)(CC)CC
C8H20N2O3
MFCD00041978
tetraethylammonium nitrate, ethanaminium, n,n,n-triethyl-, nitrate, ammonium, tetraethyl-, nitrate, ethanaminium, n,n,n-triethyl-, nitrate 1:1, tetraethylazanium nitrate, tetraethyl ammonium nitrate, tetraethylammonium nitrate w/w aqueous solution
tetraethylazanium;nitrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 217-725-5

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only