Learn More
Tetraethylammonium perchlorate, 98% (dry wt.), cont. ca 10% water, Thermo Scientific Chemicals
$119.99 - $436.45
Chemical Identifiers
CAS | 2567-83-1 |
---|---|
Molecular Formula | C8H20ClNO4 |
Molecular Weight (g/mol) | 229.70 |
MDL Number | MFCD00041977 |
InChI Key | WGHUNMFFLAMBJD-UHFFFAOYSA-M |
Synonym | tetraethylammonium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate, tetraethylazanium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate 1:1, ammonium, tetraethyl-, perchlorate, tetraethylammoniumperchlorate, acmc-1ccq7, ksc203k4p, tetraethyl ammonium perchlorate, tetraethylammoniumperchlorate, dry |
PubChem CID | 2734932 |
IUPAC Name | tetraethylazanium perchlorate |
SMILES | [O-][Cl](=O)(=O)=O.CC[N+](CC)(CC)CC |
Description
Tetraethylammonium perchlorate is used as supporting electrolyte in polarographic measurements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2567-83-1 | |
229.70 | |
WGHUNMFFLAMBJD-UHFFFAOYSA-M | |
2734932 | |
[O-][Cl](=O)(=O)=O.CC[N+](CC)(CC)CC |
C8H20ClNO4 | |
MFCD00041977 | |
tetraethylammonium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate, tetraethylazanium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate 1:1, ammonium, tetraethyl-, perchlorate, tetraethylammoniumperchlorate, acmc-1ccq7, ksc203k4p, tetraethyl ammonium perchlorate, tetraethylammoniumperchlorate, dry | |
tetraethylazanium perchlorate |
Specifications
2567-83-1 | |
Odorless | |
MFCD00041977 | |
UN1479 | |
tetraethylammonium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate, tetraethylazanium perchlorate, ethanaminium, n,n,n-triethyl-, perchlorate 1:1, ammonium, tetraethyl-, perchlorate, tetraethylammoniumperchlorate, acmc-1ccq7, ksc203k4p, tetraethyl ammonium perchlorate, tetraethylammoniumperchlorate, dry | |
WGHUNMFFLAMBJD-UHFFFAOYSA-M | |
tetraethylazanium perchlorate | |
2734932 | |
98% | |
Wet Crystals | |
(dry wt.) |
>300°C | |
C8H20ClNO4 | |
10 g | |
3638093 | |
Soluble in water,acetone,acetonitrile and dimethyl sulfoxide. | |
[O-][Cl](=O)(=O)=O.CC[N+](CC)(CC)CC | |
229.70 | |
229.71 | |
Contains ≈ 10% water | |
Tetraethylammonium perchlorate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P283-P302+P352-P304+P340-P305+P351+P338-P306+P360-P312-P332+P313-P363-P370+P378q-P371+P380+P375-P501c
H271-H315-H319-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber : 219-904-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only