missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylene glycol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B23880AP

 View more versions of this product

Catalog No. AAB23880AP


Explore available promotions

Description

Description

Tetraethylene glycol is used as a solvent to dissolve water-insoluble compounds. It is employed as a nitrocellulose solvent, a plasticizer, a natural gas desiccant, a hydraulic fluid and in lacquers and coatings. Furthermore, it is used in the synthesis of tetraethylene glycol methacrylate monomer. It acts as a chemical intermediate and acts as a solvent for the manufacturing of inks and dyes. In addition, it serves as a coupling agent in the production of textile lubricants and formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylene glycol is used as a solvent to dissolve water-insoluble compounds. It is employed as a nitrocellulose solvent, a plasticizer, a natural gas desiccant, a hydraulic fluid and in lacquers and coatings. Furthermore, it is used in the synthesis of tetraethylene glycol methacrylate monomer. It acts as a chemical intermediate and acts as a solvent for the manufacturing of inks and dyes. In addition, it serves as a coupling agent in the production of textile lubricants and formulations.

Solubility
Miscible with water, ethanol, acetone, acetic acid, glycerin, pyridine, aldehydes, carbon tetrachloride and diethyl ether. Immiscible with oil, benzene, toluene, fat and aliphatic hydrocarbons.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Tetraethylene glycol
-4°C
8.5
193°C (379°F)
C8H18O5
MFCD00002879
1634320
Miscible with water,ethanol,acetone,acetic acid,glycerin,pyridine,aldehydes,carbon tetrachloride and diethyl ether. Immiscible with oil,benzene,toluene,fat and aliphatic hydrocarbons.
C(COCCOCCOCCO)O
194.227
CHEBI:44920
99%
112-60-7
1.125
313°C to 314°C
Odorless
1.459
500 mL
tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843
UWHCKJMYHZGTIT-UHFFFAOYSA-N
2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
8200
194.23

Chemical Identifiers

112-60-7
194.227
UWHCKJMYHZGTIT-UHFFFAOYSA-N
8200
2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethanol
C8H18O5
MFCD00002879
tetraethylene glycol, tetraglycol, 2,2'-oxybis ethane-2,1-diyl bis oxy diethanol, hi-dry, carbitol, diethyl, 3,6,9-trioxaundecane-1,11-diol, bis 2-2-hydroxyethoxy ethyl ether, ethanol, 2,2'-oxybis 2,1-ethanediyloxy bis, tetra ethylene glycol, hsdb 843
CHEBI:44920
C(COCCOCCOCCO)O
Safety and Handling

Safety and Handling

P201-P202-P262-P264b-P280-P302+P352-P305+P351+P338-P308+P313-P314-P332+P313-P362-P501c

H315-H320-H341-H360-H373

EINECSNumber : 203-989-9

RTECSNumber : XC2100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only