Learn More
Tetraethylenepentamine, tech., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 138112500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetraethylenepentamine | |
112-57-2 | |
0.9900g/mL | |
139°C | |
Glass bottle | |
1.5055 | |
MFCD00008168 | |
0.99 | |
Solubility in water: soluble | |
NCCNCCNCCNCCN | |
189.31 | |
CHEBI:49798 | |
≥95.0% | |
Liquid |
tech. | |
-40°C | |
340°C | |
Authentic | |
C8H23N5 | |
HN(CH2CH2NHCH2CH2NH2)2 | |
250 g | |
tetraethylenepentamine, tetren, 1,4,7,10,13-pentaazatridecane, tetraethylene pentamine, tetraethylpentylamine, 1,11-diamino-3,6,9-triazaundecane, 3,6,9-triazaundecamethylenediamine, 3,6,9-triazaundecane-1,11-diamine, deh 26, unii-yzd1c9kq28 | |
FAGUFWYHJQFNRV-UHFFFAOYSA-N | |
N'-[2-[2-(2-aminoethylamino)ethylamino]ethyl]ethane-1,2-diamine | |
8197 | |
189.3 | |
Technical |
Chemical Identifiers
112-57-2 | |
189.31 | |
FAGUFWYHJQFNRV-UHFFFAOYSA-N | |
8197 | |
N'-[2-[2-(2-aminoethylamino)ethylamino]ethyl]ethane-1,2-diamine |
C8H23N5 | |
MFCD00008168 | |
tetraethylenepentamine, tetren, 1,4,7,10,13-pentaazatridecane, tetraethylene pentamine, tetraethylpentylamine, 1,11-diamino-3,6,9-triazaundecane, 3,6,9-triazaundecamethylenediamine, 3,6,9-triazaundecane-1,11-diamine, deh 26, unii-yzd1c9kq28 | |
CHEBI:49798 | |
NCCNCCNCCNCCN |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Danger
EINECSNumber : 203-986-2
RTECSNumber : KH8585000
TSCA : TSCA
RUO – Research Use Only