missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraethylenepentamine, tech., Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 138115000

 View more versions of this product

Catalog No. AC138115000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetraethylenepentamine
112-57-2
0.9900g/mL
139°C
Glass bottle
1.5055
MFCD00008168
0.99
Solubility in water: soluble
NCCNCCNCCNCCN
189.31
CHEBI:49798
≥95.0%
Liquid
tech.
-40°C
340°C
Authentic
C8H23N5
HN(CH2CH2NHCH2CH2NH2)2
500 g
tetraethylenepentamine, tetren, 1,4,7,10,13-pentaazatridecane, tetraethylene pentamine, tetraethylpentylamine, 1,11-diamino-3,6,9-triazaundecane, 3,6,9-triazaundecamethylenediamine, 3,6,9-triazaundecane-1,11-diamine, deh 26, unii-yzd1c9kq28
FAGUFWYHJQFNRV-UHFFFAOYSA-N
N'-[2-[2-(2-aminoethylamino)ethylamino]ethyl]ethane-1,2-diamine
8197
189.3
Technical

Chemical Identifiers

112-57-2
189.31
FAGUFWYHJQFNRV-UHFFFAOYSA-N
8197
N'-[2-[2-(2-aminoethylamino)ethylamino]ethyl]ethane-1,2-diamine
C8H23N5
MFCD00008168
tetraethylenepentamine, tetren, 1,4,7,10,13-pentaazatridecane, tetraethylene pentamine, tetraethylpentylamine, 1,11-diamino-3,6,9-triazaundecane, 3,6,9-triazaundecamethylenediamine, 3,6,9-triazaundecane-1,11-diamine, deh 26, unii-yzd1c9kq28
CHEBI:49798
NCCNCCNCCNCCN
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously wit

GHS Signal Word: Danger

EINECSNumber : 203-986-2

RTECSNumber : KH8585000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only