Learn More
Tetraethyltin, 98%, Thermo Scientific Chemicals
$1291.45 - $1291.45
Chemical Identifiers
CAS | 597-64-8 |
---|---|
Molecular Formula | C8H20Sn |
Molecular Weight (g/mol) | 234.96 |
MDL Number | MFCD00009020 |
InChI Key | RWWNQEOPUOCKGR-UHFFFAOYSA-N |
Synonym | tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa |
PubChem CID | 11704 |
IUPAC Name | tetraethylstannane |
SMILES | CC[Sn](CC)(CC)CC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AA7114418
|
Thermo Scientific Chemicals
07114418 |
50 g |
Each for $1,291.45
|
|
|||||
Description
Tetraethyltin is used in the electronics industry. Used as catalyst for olefin polymerization and to produce other organotin compounds. Also used as a preservative for wood, textiles, paper, and leather.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
597-64-8 | |
234.96 | |
RWWNQEOPUOCKGR-UHFFFAOYSA-N | |
11704 | |
CC[Sn](CC)(CC)CC |
C8H20Sn | |
MFCD00009020 | |
tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa | |
tetraethylstannane |
Specifications
597-64-8 | |
1.187 g/mL | |
53°C (128°F) | |
C8H20Sn | |
MFCD00009020 | |
UN3384 | |
Insoluble in water; Soluble in alcohol and ether | |
CC[Sn](CC)(CC)CC | |
234.96 | |
234.94 | |
Liquid |
-112°C | |
181°C | |
Characteristic | |
1.473 | |
50 g | |
tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa | |
RWWNQEOPUOCKGR-UHFFFAOYSA-N | |
tetraethylstannane | |
11704 | |
98% | |
Tetraethyltin |
Safety and Handling
GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P310-P330-P363-P370+P378q-P501c
H226-H300+H310+H330
DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
EINECSNumber : 209-906-2
RTECSNumber : WH8625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only