Learn More
Tetrafluoroboric acid, 48% min w/w aq. soln., Thermo Scientific Chemicals
$100.81 - $378.01
Chemical Identifiers
CAS | 16872-11-0 |
---|---|
MDL Number | MFCD00011345 |
Synonym | fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA1148418
|
Thermo Scientific Chemicals
01148418 |
50 g |
Each for $100.81
|
|
||||||
AA1148436
|
Thermo Scientific Chemicals
01148436 |
500 g |
Each for $139.70
|
|
||||||
AA1148457
|
Thermo Scientific Chemicals
01148457 |
4 x 500 g |
Each for $378.01
|
|
||||||
Description
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and treat the surface of substrates. It is used as radiotracers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
16872-11-0 | |
MFCD00011345 | |
fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq |
Specifications
16872-11-0 | |
1.4 | |
Liquid | |
fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq | |
87.81 |
130°C (decomposition) | |
50 g | |
MFCD00011345 | |
Miscible with water. | |
Tetrafluoroboric Acid |
Safety and Handling
P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H360FD
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 240-898-3
RTECSNumber : ED2685000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only