missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrafluoroisophthalonitrile 98.0+%, TCI America™
$162.17 - $430.60
Chemical Identifiers
| CAS | 2377-81-3 |
|---|---|
| Molecular Formula | C8F4N2 |
| Molecular Weight (g/mol) | 200.10 |
| MDL Number | MFCD00013290 |
| InChI Key | WVHMPQKZPHOCRD-UHFFFAOYSA-N |
| Synonym | tetrafluoroisophthalonitrile, 2,4,5,6-tetrafluoroisophthalonitrile, perfluoroisophthalonitrile, tetrafluorobenzene-1,3-dicarbonitrile, 1,3-benzenedicarbonitrile, 2,4,5,6-tetrafluoro, tetrafluoroisopthalonitrile, m-dicyanotetrafluorobenzene, acmc-209g5p, 1,3-dicyanoperfluorobenzene |
| PubChem CID | 608182 |
| IUPAC Name | tetrafluorobenzene-1,3-dicarbonitrile |
| SMILES | FC1=C(F)C(C#N)=C(F)C(C#N)=C1F |
Chemical Identifiers
| 2377-81-3 | |
| 200.10 | |
| WVHMPQKZPHOCRD-UHFFFAOYSA-N | |
| 608182 | |
| FC1=C(F)C(C#N)=C(F)C(C#N)=C1F |
| C8F4N2 | |
| MFCD00013290 | |
| tetrafluoroisophthalonitrile, 2,4,5,6-tetrafluoroisophthalonitrile, perfluoroisophthalonitrile, tetrafluorobenzene-1,3-dicarbonitrile, 1,3-benzenedicarbonitrile, 2,4,5,6-tetrafluoro, tetrafluoroisopthalonitrile, m-dicyanotetrafluorobenzene, acmc-209g5p, 1,3-dicyanoperfluorobenzene | |
| tetrafluorobenzene-1,3-dicarbonitrile |
Specifications
| 2377-81-3 | |
| White-Yellow | |
| MFCD00013290 | |
| tetrafluoroisophthalonitrile, 2,4,5,6-tetrafluoroisophthalonitrile, perfluoroisophthalonitrile, tetrafluorobenzene-1,3-dicarbonitrile, 1,3-benzenedicarbonitrile, 2,4,5,6-tetrafluoro, tetrafluoroisopthalonitrile, m-dicyanotetrafluorobenzene, acmc-209g5p, 1,3-dicyanoperfluorobenzene | |
| FC1=C(F)C(C#N)=C(F)C(C#N)=C1F | |
| 200.10 | |
| 200.10 | |
| Crystalline Powder |
| 76°C | |
| C8F4N2 | |
| 5 g | |
| WVHMPQKZPHOCRD-UHFFFAOYSA-N | |
| tetrafluorobenzene-1,3-dicarbonitrile | |
| 608182 | |
| ≥98.0% (GC) | |
| Tetrafluoroisophthalonitrile |
Safety and Handling
EINECSNumber : (3)-3882
TSCA : No