missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2-furoic acid, 98+%

CAS: 16874-33-2 | C5H8O3 | 116.12 g/mol

Supplier:  Thermo Scientific Chemicals 263140250

 View more versions of this product

Catalog No. AC263140250


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetrahydro-2-furoic acid
16874-33-2
99.0
1.1700g/mL
25 g
Glass bottle
1.4590 to 1.4610
1.17
MFCD00022473
Solubility in water: soluble
C1CC(OC1)C(=O)O
116.12
116.12
99+%
Yellow to Brown
100.0
128.0°C to 129.0°C (13.0 mmHg)
Authentic
Liquid
98% min. (GC)
C5H8O3
2-tetrahydrofuroic acid, tetrahydrofuran-2-carboxylic acid, tetrahydro-2-furoic acid, tetrahydro-2-furancarboxylic acid, 2-furancarboxylic acid, tetrahydro, furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, thfc, tetrahydrofuroic acid, pubchem7088
UJJLJRQIPMGXEZ-UHFFFAOYSA-N
oxolane-2-carboxylic acid
86079
98+%

Chemical Identifiers

16874-33-2
116.12
UJJLJRQIPMGXEZ-UHFFFAOYSA-N
86079
C1CC(OC1)C(=O)O
C5H8O3
MFCD00022473
2-tetrahydrofuroic acid, tetrahydrofuran-2-carboxylic acid, tetrahydro-2-furoic acid, tetrahydro-2-furancarboxylic acid, 2-furancarboxylic acid, tetrahydro, furancarboxylic acid, tetrahydro, 2-furoic acid, tetrahydro, thfc, tetrahydrofuroic acid, pubchem7088
oxolane-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only