missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydro-2H-pyran-4-carbonyl chloride, ≥97%, Thermo Scientific™
$145.24 - $145.24
Chemical Identifiers
| CAS | 40191-32-0 |
|---|---|
| Molecular Formula | C6H9ClO2 |
| Molecular Weight (g/mol) | 148.59 |
| MDL Number | MFCD06200863 |
| InChI Key | RYGUCYSSMOFTSH-UHFFFAOYSA-N |
| Synonym | tetrahydro-2h-pyran-4-carbonyl chloride, 2h-pyran-4-carbonyl chloride, tetrahydro, tetrahydro-2h-pyran-4-carbonylchloride, pubchem12437, ksc235m5r, tetrahydropyran-4-carbonyl chloride, tetrahydro-pyran-4-carbonyl chloride, 4-chloroformyl tetrahydro-2h-pyran, tetrahydropyran-4-carboxylic chloride, 4-chlorocarbonyl tetrahydro-2h-pyran |
| PubChem CID | 2795505 |
| IUPAC Name | oxane-4-carbonyl chloride |
| SMILES | ClC(=O)C1CCOCC1 |
Chemical Identifiers
| 40191-32-0 | |
| 148.59 | |
| RYGUCYSSMOFTSH-UHFFFAOYSA-N | |
| 2795505 | |
| ClC(=O)C1CCOCC1 |
| C6H9ClO2 | |
| MFCD06200863 | |
| tetrahydro-2h-pyran-4-carbonyl chloride, 2h-pyran-4-carbonyl chloride, tetrahydro, tetrahydro-2h-pyran-4-carbonylchloride, pubchem12437, ksc235m5r, tetrahydropyran-4-carbonyl chloride, tetrahydro-pyran-4-carbonyl chloride, 4-chloroformyl tetrahydro-2h-pyran, tetrahydropyran-4-carboxylic chloride, 4-chlorocarbonyl tetrahydro-2h-pyran | |
| oxane-4-carbonyl chloride |
Specifications
| 40191-32-0 | |
| 100.0 | |
| Amber Glass Bottle | |
| MFCD06200863 | |
| tetrahydro-2h-pyran-4-carbonyl chloride, 2h-pyran-4-carbonyl chloride, tetrahydro, tetrahydro-2h-pyran-4-carbonylchloride, pubchem12437, ksc235m5r, tetrahydropyran-4-carbonyl chloride, tetrahydro-pyran-4-carbonyl chloride, 4-chloroformyl tetrahydro-2h-pyran, tetrahydropyran-4-carboxylic chloride, 4-chlorocarbonyl tetrahydro-2h-pyran | |
| ClC(=O)C1CCOCC1 | |
| 148.59 | |
| 148.59 | |
| Tetrahydro-2H-pyran-4-carbonyl chloride |
| 94.5 | |
| 95% | |
| C6H9ClO2 | |
| 1 g | |
| RYGUCYSSMOFTSH-UHFFFAOYSA-N | |
| oxane-4-carbonyl chloride | |
| 2795505 | |
| 95% |
Safety and Handling
GHS P Statement Causes severe skin burns and eye damage. Reacts violently with water. Contact with water liberates toxic gas.
GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair):
Danger