missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-β-carboline, 98%, Thermo Scientific™

Chemical Identifiers

CAS 16502-01-5
Molecular Formula C11H13ClN2
Molecular Weight (g/mol) 208.69
MDL Number MFCD00004954
InChI Key PHLJRXUBLWEPCM-UHFFFAOYSA-N
Synonym tryptoline, 2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, noreleagnine, 1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 1,2,3,4-tetrahydro-beta-carboline, tetrahydro-beta-carboline, thbc, tetrahydronorharman, unii-65027tmi0h, 1h,2h,3h,4h,9h-pyrido 3,4-b indole
PubChem CID 107838
IUPAC Name 2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
SMILES [H+].[Cl-].C1CC2=C(CN1)NC1=CC=CC=C21
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC427580010
SDS View Documents
Thermo Scientific Chemicals
427580010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC427580050
SDS View Documents
Thermo Scientific Chemicals
427580050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

16502-01-5
208.69
PHLJRXUBLWEPCM-UHFFFAOYSA-N
107838
[H+].[Cl-].C1CC2=C(CN1)NC1=CC=CC=C21
C11H13ClN2
MFCD00004954
tryptoline, 2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, noreleagnine, 1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 1,2,3,4-tetrahydro-beta-carboline, tetrahydro-beta-carboline, thbc, tetrahydronorharman, unii-65027tmi0h, 1h,2h,3h,4h,9h-pyrido 3,4-b indole
2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole

Specifications

16502-01-5
97.5% min. (HPLC)
C11H13ClN2
1g
PHLJRXUBLWEPCM-UHFFFAOYSA-N
2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole
107838
98%
Authentic
Glass bottle
MFCD00004954
tryptoline, 2,3,4,9-tetrahydro-1h-pyrido 3,4-b indole, noreleagnine, 1,2,3,4-tetrahydro-9h-pyrido 3,4-b indole, 1,2,3,4-tetrahydro-beta-carboline, tetrahydro-beta-carboline, thbc, tetrahydronorharman, unii-65027tmi0h, 1h,2h,3h,4h,9h-pyrido 3,4-b indole
[H+].[Cl-].C1CC2=C(CN1)NC1=CC=CC=C21
208.69
172.23
Tetrahydro-β-carboline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

SDS
Documents

Documents

RUO – Research Use Only