missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrahydrofuran-3-sulfonyl chloride, Thermo Scientific™
Supplier: Thermo Scientific Chemicals CC76003DE
Specifications
Tetrahydrofuran-3-sulfonyl chloride | |
Amber Glass Bottle | |
5g | |
MVZIUOJNVVIOEY-UHFFFAOYSA-N | |
oxolane-3-sulfonyl chloride | |
49762850 | |
97% |
1207346-29-9 | |
C4H7ClO3S | |
tetrahydrofuran-3-sulfonyl chloride, chlorooxolan-3-ylsulfone | |
C1COCC1S(=O)(=O)Cl | |
170.607 | |
170.61 |
Safety and Handling
GHS Signal Word: Danger