Learn More
Tetrahydrofuran, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
$71.25 - $410.54
Chemical Identifiers
CAS | 109-99-9 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00005356 |
InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
Synonym | tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano |
PubChem CID | 8028 |
ChEBI | CHEBI:26911 |
IUPAC Name | oxolane |
SMILES | C1CCOC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC348451000
|
Thermo Scientific Chemicals
348451000 |
100 mL |
Each for $71.25
|
|
|||||
AC348450010
|
Thermo Scientific Chemicals
348450010 |
1 L |
Each for $204.01
|
|
|||||
AC348450025
|
Thermo Scientific Chemicals
348450025 |
2.5 L |
Each for $410.54
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-99-9 | |
72.11 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
8028 | |
oxolane |
C4H8O | |
MFCD00005356 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
CHEBI:26911 | |
C1CCOC1 |
Specifications
-108.4°C | |
Colorless | |
66.0°C | |
−21°C | |
1.407 | |
0.88 | |
MFCD00005356 | |
0.005% Max. (Coulometric) | |
01,1140; 02,398; 03,278; 05,649; 06,570 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
oxolane | |
8028 | |
72.11 | |
Extra Dry over Molecular Sieve |
109-99-9,128-37-0 | |
0.8800g/mL | |
100 mL | |
Liquid | |
99.5% | |
C4H8O | |
0.55 mPa.s (20°C) | |
17, 10 | |
15, 9356 | |
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone | |
C1CCOC1 | |
72.11 | |
CHEBI:26911 | |
99.5% | |
Tetrahydrofuran, Water <50 ppm, stabilized |
Safety and Handling
GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed.
Danger
RUO – Research Use Only