Learn More
Tetrahydrofuran (HPLC), Fisher Chemical™
HPLC mobile phase for analytical and preparative separations
Supplier: Fisher Chemical T4254
Specifications
7 to 8 | |
-108.4°C | |
Pass Test, 1.0 AU max. at 210nm, 0.6 AU max. at 215nm, 0.3 AU max. at 230nm, 0.1 AU max. at 254nm | |
0.880g/cm³ | |
4 L | |
Pass Test | |
Liquid | |
5ppm max. | |
C4H8O | |
MFCD00005356 | |
0.55 cP at 20°C | |
0.02% max. | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
oxolane | |
8028 | |
72.11 | |
HPLC |
Tetrahydrofuran | |
109-99-9 | |
Colorless | |
66°C | |
â21°C(-69.6°F) | |
Glass Bottle | |
Mobile phase for HPLC applications | |
99.9% min by GC | |
CH2(CH2)2CH2O | |
200mbar at 20°C | |
2056 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
C1CCOC1 | |
72.11 | |
CHEBI:26911 | |
≥99.9% |
Chemical Identifiers
109-99-9 | |
72.11 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
8028 | |
oxolane |
C4H8O | |
MFCD00005356 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
CHEBI:26911 | |
C1CCOC1 |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.
NFPA
Health:2
Flammability:3
Instability:1
DOTInformation : DOT Class 3, : Flammable Liquid