missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, Optima™ for HPLC, Fisher Chemical™

High purity solvent for HPLC

Supplier:  Thermo Fisher Scientific T4274

 View more versions of this product

Catalog No. T427-4



Only null left
Explore available promotions

Description

Description

  • Screened for fluorescing and UV-absorbing contaminants to reduce interferences for smoother baselines
  • Also meets ACS specifications
  • Intended for research use only
Specifications

Specifications

7 to 8
-108.4°C
1.0 AU max. at 210nm, 0.6 AU max. at 215nm, 0.3 AU max. at 230nm, 0.1 AU max. at 254nm
0.880g/cm³
4 L
Amber Soda Lime Glass
High purity mobile phase for HPLC
5ppm max.
C4H8O
200mbar at 20°C
2056
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
C1CCOC1
72.11
CHEBI:26911
≥99.9%
Tetrahydrofuran
109-99-9
Colorless
66°C
Pass Test
Liquid
1.4040 to 1.4070 (at 25°C)
99.9% min by GC
MFCD00005356
0.55 cP at 20°C
0.02% max.
WYURNTSHIVDZCO-UHFFFAOYSA-N
oxolane
8028
72.11
OPTIMA

Chemical Identifiers

109-99-9
72.11
WYURNTSHIVDZCO-UHFFFAOYSA-N
8028
oxolane
C4H8O
MFCD00005356
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
CHEBI:26911
C1CCOC1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Possible cancer hazard May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. May cause central nervous system effects. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. If peroxide formation is suspected, do not open or move container. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:1

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Promotions

Promotions