missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrahydropyran-4-ylacetic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC67401DA

 View more versions of this product

Catalog No. CC67401DA


Description

Description

Specifications

Specifications

Tetrahydropyran-4-ylacetic acid
96.5
97%
C7H11O3
1 g
PBXYNWPYMVWJAH-UHFFFAOYSA-M
2-(oxan-4-yl)acetic acid
2773575
97%
85064-61-5
100.0
Amber Glass Bottle
MFCD01631204
tetrahydropyranyl-4-acetic acid, tetrahydropyran-4-yl-acetic acid, 2-oxan-4-yl acetic acid, 2-tetrahydro-2h-pyran-4-yl acetic acid, tetrahydropyran-4-ylacetic acid, oxan-4-ylacetic acid, tetrahydro-2h-pyran-4-ylacetic acid, 2h-pyran-4-acetic acid, tetrahydro, 2-4-tetrahydropyranyl acetic acid, tetrahydropyran-4-acetic acid
[O-]C(=O)CC1CCOCC1
143.16
144.17

Chemical Identifiers

85064-61-5
143.16
PBXYNWPYMVWJAH-UHFFFAOYSA-M
2773575
[O-]C(=O)CC1CCOCC1
C7H11O3
MFCD01631204
tetrahydropyranyl-4-acetic acid, tetrahydropyran-4-yl-acetic acid, 2-oxan-4-yl acetic acid, 2-tetrahydro-2h-pyran-4-yl acetic acid, tetrahydropyran-4-ylacetic acid, oxan-4-ylacetic acid, tetrahydro-2h-pyran-4-ylacetic acid, 2h-pyran-4-acetic acid, tetrahydro, 2-4-tetrahydropyranyl acetic acid, tetrahydropyran-4-acetic acid
2-(oxan-4-yl)acetic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions