Learn More
Tetrahydrothiophene 1-oxide, 97%, Thermo Scientific Chemicals
$81.81 - $314.10
Chemical Identifiers
CAS | 1600-44-8 |
---|---|
Molecular Formula | C4H8OS |
Molecular Weight (g/mol) | 104.167 |
MDL Number | MFCD00005477 |
InChI Key | ISXOBTBCNRIIQO-UHFFFAOYSA-N |
Synonym | tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide |
PubChem CID | 1128 |
IUPAC Name | thiolane 1-oxide |
SMILES | C1CCS(=O)C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1750209
|
Thermo Scientific Chemicals
A1750209 |
10 g |
Each for $81.81
|
|
|||||
AAA1750218
|
Thermo Scientific Chemicals
A1750218 |
50 g |
Each for $314.10
|
|
|||||
Description
In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIn a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.
Solubility
Fully miscible in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
1600-44-8 | |
104.167 | |
ISXOBTBCNRIIQO-UHFFFAOYSA-N | |
1128 | |
C1CCS(=O)C1 |
C4H8OS | |
MFCD00005477 | |
tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide | |
thiolane 1-oxide |
Specifications
1600-44-8 | |
235°C to 237°C | |
C4H8OS | |
MFCD00005477 | |
105274 | |
Fully miscible in water. | |
C1CCS(=O)C1 | |
104.167 | |
104.17 | |
Tetrahydrothiophene 1-oxide |
1.173 | |
112°C (233°F) | |
1.52 | |
10 g | |
tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide | |
ISXOBTBCNRIIQO-UHFFFAOYSA-N | |
thiolane 1-oxide | |
1128 | |
97% |
Safety and Handling
EINECSNumber : 216-493-2
RTECSNumber : XN0830000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only