Learn More
Tetraisopropyl methylenediphosphonate, 98%, Thermo Scientific Chemicals
$107.58 - $1,488.33
Chemical Identifiers
CAS | 1660-95-3 |
---|---|
Molecular Formula | C13H30O6P2 |
Molecular Weight (g/mol) | 344.325 |
MDL Number | MFCD00015021 |
InChI Key | ODTQUKVFOLFLIQ-UHFFFAOYSA-N |
Synonym | tetraisopropyl methylenediphosphonate, phosphonic acid, methylenebis-, tetrakis 1-methylethyl ester, tetraisopropyl methylenebisphosphonate, unii-k46y5v9u61, tetraisopropyl methylenebis phosphonate, methylenediphosphonic acid tetraisopropyl ester, tetra-i-propylmethylenediphosphonate, 2-di propan-2-yloxy phosphorylmethyl-propan-2-yloxyphosphoryl oxypropane, phosphonic acid, p,p'-methylenebis-, p,p,p',p'-tetrakis 1-methylethyl ester, dsstox_cid_24847 |
PubChem CID | 74266 |
IUPAC Name | 2-[di(propan-2-yloxy)phosphorylmethyl-propan-2-yloxyphosphoryl]oxypropane |
SMILES | CC(C)OP(=O)(CP(=O)(OC(C)C)OC(C)C)OC(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1348409
|
Thermo Scientific Chemicals
A1348409 |
10 g |
N/A
|
|
||||||
AAA1348418
|
Thermo Scientific Chemicals
A1348418 |
50 g |
N/A
|
|
||||||
AAA1348430
|
Thermo Scientific Chemicals
A1348430 |
250 g |
N/A
|
|
||||||
Description
Tetraisopropyl methylenediphosphonate is used in the preparation of AZT 5â-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetraisopropyl methylenediphosphonate is used in the preparation of AZT 5â-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Chemical Identifiers
1660-95-3 | |
344.325 | |
ODTQUKVFOLFLIQ-UHFFFAOYSA-N | |
74266 | |
CC(C)OP(=O)(CP(=O)(OC(C)C)OC(C)C)OC(C)C |
C13H30O6P2 | |
MFCD00015021 | |
tetraisopropyl methylenediphosphonate, phosphonic acid, methylenebis-, tetrakis 1-methylethyl ester, tetraisopropyl methylenebisphosphonate, unii-k46y5v9u61, tetraisopropyl methylenebis phosphonate, methylenediphosphonic acid tetraisopropyl ester, tetra-i-propylmethylenediphosphonate, 2-di propan-2-yloxy phosphorylmethyl-propan-2-yloxyphosphoryl oxypropane, phosphonic acid, p,p'-methylenebis-, p,p,p',p'-tetrakis 1-methylethyl ester, dsstox_cid_24847 | |
2-[di(propan-2-yloxy)phosphorylmethyl-propan-2-yloxyphosphoryl]oxypropane |
Specifications
1660-95-3 | |
122°C to 124°C (1.5 mmHg) | |
C13H30O6P2 | |
MFCD00015021 | |
1080180 | |
Insoluble in water. | |
CC(C)OP(=O)(CP(=O)(OC(C)C)OC(C)C)OC(C)C | |
344.325 | |
344.33 | |
Tetraisopropyl methylenediphosphonate |
1.08 | |
160°C (320°F) | |
1.434 | |
10 g | |
tetraisopropyl methylenediphosphonate, phosphonic acid, methylenebis-, tetrakis 1-methylethyl ester, tetraisopropyl methylenebisphosphonate, unii-k46y5v9u61, tetraisopropyl methylenebis phosphonate, methylenediphosphonic acid tetraisopropyl ester, tetra-i-propylmethylenediphosphonate, 2-di propan-2-yloxy phosphorylmethyl-propan-2-yloxyphosphoryl oxypropane, phosphonic acid, p,p'-methylenebis-, p,p,p',p'-tetrakis 1-methylethyl ester, dsstox_cid_24847 | |
ODTQUKVFOLFLIQ-UHFFFAOYSA-N | |
2-[di(propan-2-yloxy)phosphorylmethyl-propan-2-yloxyphosphoryl]oxypropane | |
74266 | |
98% |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
EINECSNumber : 216-765-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only