missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), 99.9%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 3954803

 View more versions of this product

Catalog No. AA3954803


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV)
318°C to 320°C
C44H80O8Zr
1 g
ANBXKEDJQLYCLX-UHFFFAOYSA-N
5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium
50919870
99.9%
18865-74-2
180°C (0.1 mmHg)(sublimation)
MFCD00145380,MFCD00145380
tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium, zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate, tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv, zr tmhd 4, zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, zirconium tetrakis dipivaloylmethanate, zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate, 4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one
[Zr].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
828.34
824.3

Chemical Identifiers

18865-74-2
828.34
ANBXKEDJQLYCLX-UHFFFAOYSA-N
50919870
[Zr].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C
C44H80O8Zr
MFCD00145380,MFCD00145380
tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium, zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate, tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv, zr tmhd 4, zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, zirconium tetrakis dipivaloylmethanate, zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate, 4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one
5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions