missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetrakis(acetonitrile)copper(I) Hexafluorophosphate 97.0+%, TCI America™
Supplier: TCI America T26655G
Specifications
Tetrakis(acetonitrile)copper(I) Hexafluorophosphate | |
160°C | |
C8H12CuF6N4P | |
5 g | |
CC#N.CC#N.CC#N.CC#N.F[P-](F)(F)(F)(F)F.[Cu+] | |
372.722 | |
372.72 | |
Crystalline Powder |
64443-05-6 | |
White | |
MFCD00064810 | |
GNQXUMGHBSAQBV-UHFFFAOYSA-N | |
acetonitrile;copper(1+);hexafluorophosphate | |
11068737 | |
≥97.0% (T) |
Chemical Identifiers
64443-05-6 | |
372.722 | |
GNQXUMGHBSAQBV-UHFFFAOYSA-N | |
acetonitrile;copper(1+);hexafluorophosphate |
C8H12CuF6N4P | |
MFCD00064810 | |
11068737 | |
CC#N.CC#N.CC#N.CC#N.F[P-](F)(F)(F)(F)F.[Cu+] |
Safety and Handling
TSCA : No
Recommended Storage : Freezer