missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetrakis(triphenylphosphine)palladium(0) 97.0+%, TCI America™

Supplier:  TCI America T135025G

 View more versions of this product

Catalog No. T135025G


Explore available promotions

Description

Description

Specifications

Specifications

Tetrakis(triphenylphosphine)palladium(0)
105°C
MFCD00010012
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4
[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
1155.59
1155.59
Crystalline Powder
14221-01-3
C72H60P4Pd
25 g
NFHFRUOZVGFOOS-UHFFFAOYSA-N
tetrakis(triphenylphosphane) palladium
11979704
≥97.0% (T)

Chemical Identifiers

14221-01-3
1155.59
NFHFRUOZVGFOOS-UHFFFAOYSA-N
11979704
[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
C72H60P4Pd
MFCD00010012
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4
tetrakis(triphenylphosphane) palladium
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions