Learn More
Tetramethyl methylenediphosphonate, 98+%, Thermo Scientific Chemicals
$228.48 - $2948.67
Chemical Identifiers
CAS | 16001-93-7 |
---|---|
Molecular Formula | C5H14O6P2 |
Molecular Weight (g/mol) | 232.109 |
MDL Number | MFCD00014884 |
InChI Key | XAVFZUKFLWOSOS-UHFFFAOYSA-N |
Synonym | tetramethyl methylenediphosphonate, bis dimethoxyphosphoryl methane, tetramethyl methylenebis phosphonate, phosphonic acid, methylenebis-, tetramethyl ester, acmc-20ajjd, tetramethyl methanebisphosphonate, tetramethyl-methylenediphosphonate, tetramethyl methylenebisphosphonate, methylenebis phosphonic acid dimethyl ester, methylenediphosphonic acid tetramethyl ester |
PubChem CID | 519206 |
IUPAC Name | bis(dimethoxyphosphoryl)methane |
SMILES | COP(=O)(CP(=O)(OC)OC)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1344106
|
Thermo Scientific Chemicals
A1344106 |
5 g |
Each for $228.48
|
|
|||||
AAA1344114
|
Thermo Scientific Chemicals
A1344114 |
25 g |
Each for $870.88
|
|
|||||
AAA1344122
|
Thermo Scientific Chemicals
A1344122 |
100 g |
Each for $2,948.67
|
|
|||||
Description
Tetramethyl methylenediphosphonate is used in the preparation of phosphonate analogue of ribose-1-phosphate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetramethyl methylenediphosphonate is used in the preparation of phosphonate analogue of ribose-1-phosphate.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
16001-93-7 | |
232.109 | |
XAVFZUKFLWOSOS-UHFFFAOYSA-N | |
519206 | |
COP(=O)(CP(=O)(OC)OC)OC |
C5H14O6P2 | |
MFCD00014884 | |
tetramethyl methylenediphosphonate, bis dimethoxyphosphoryl methane, tetramethyl methylenebis phosphonate, phosphonic acid, methylenebis-, tetramethyl ester, acmc-20ajjd, tetramethyl methanebisphosphonate, tetramethyl-methylenediphosphonate, tetramethyl methylenebisphosphonate, methylenebis phosphonic acid dimethyl ester, methylenediphosphonic acid tetramethyl ester | |
bis(dimethoxyphosphoryl)methane |
Specifications
16001-93-7 | |
C5H14O6P2 | |
MFCD00014884 | |
1874756 | |
Miscible with water. | |
COP(=O)(CP(=O)(OC)OC)OC | |
232.109 | |
232.11 | |
Tetramethyl methylenediphosphonate |
152°C to 154°C (2.5 mmHg) | |
1.455 | |
5 g | |
tetramethyl methylenediphosphonate, bis dimethoxyphosphoryl methane, tetramethyl methylenebis phosphonate, phosphonic acid, methylenebis-, tetramethyl ester, acmc-20ajjd, tetramethyl methanebisphosphonate, tetramethyl-methylenediphosphonate, tetramethyl methylenebisphosphonate, methylenebis phosphonic acid dimethyl ester, methylenediphosphonic acid tetramethyl ester | |
XAVFZUKFLWOSOS-UHFFFAOYSA-N | |
bis(dimethoxyphosphoryl)methane | |
519206 | |
≥98% |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only