missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethyl orthocarbonate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6661706

 View more versions of this product

Catalog No. AAH6661706


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tetramethyl orthocarbonate
-5°C
1.017 g/mL
22°C (72°F)
1.385
MFCD00008473
UN3272
Moisture sensitive
AHJWSRRHTXRLAQ-UHFFFAOYSA-N
tetramethoxymethane
74613
95%
1850-14-2
Colorless
114°C
C5H12O4
C(OCH3)4
5 g
1737682
tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc
COC(OC)(OC)OC
136.147
136.15
Liquid

Chemical Identifiers

1850-14-2
136.147
AHJWSRRHTXRLAQ-UHFFFAOYSA-N
74613
COC(OC)(OC)OC
C5H12O4
MFCD00008473
tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc
tetramethoxymethane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 217-438-5

TSCA : No

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only