Learn More
Tetramethylammonium hydroxide, 25% w/w aq. soln., Electronic Grade, 99.9999% (metals basis), Thermo Scientific Chemicals
$106.47 - $2693.62
Chemical Identifiers
CAS | 75-59-2 |
---|---|
Molecular Formula | C4H13NO |
Molecular Weight (g/mol) | 91.154 |
MDL Number | MFCD00008280 |
InChI Key | WGTYBPLFGIVFAS-UHFFFAOYSA-M |
Synonym | tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide |
PubChem CID | 60966 |
IUPAC Name | tetramethylazanium;hydroxide |
SMILES | C[N+](C)(C)C.[OH-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AA20932AK
|
Thermo Scientific Chemicals
020932AK |
250 mL |
Each for $106.47
|
|
||||||
AA20932K2
|
Thermo Scientific Chemicals
020932K2 |
1 L |
Each for $306.79
|
|
||||||
AA20932LV
|
Thermo Scientific Chemicals
020932LV |
1 gal. |
Each for $734.12
|
|
||||||
AA20932LU
|
Thermo Scientific Chemicals
020932LU |
4 x 1 gal. |
Each for $2,693.62
|
|
||||||
Description
Tetramethylammonium hydroxide is used to produce tetramethyl-ammonium azide. It is used as an anisotropic etchant of silicon, basic solvent in the development of acidic photoresist in photolithography process, as surfactant in the synthesis of ferrofluid and as a polarographic reagent. It finds application in the production of organic silicon, where it is used for computer silicon wafer surface brightener and cleaning agent. It is also involved in the purification of some metallic elements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-59-2 | |
91.154 | |
WGTYBPLFGIVFAS-UHFFFAOYSA-M | |
60966 | |
C[N+](C)(C)C.[OH-] |
C4H13NO | |
MFCD00008280 | |
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide | |
tetramethylazanium;hydroxide |
Specifications
75-59-2 | |
1.016 g/mL | |
C4H13NO | |
(CH3)4NOH | |
250 mL | |
3558708 | |
14,9224 | |
Miscible with water. | |
C[N+](C)(C)C.[OH-] | |
91.154 | |
91.15 | |
Electronic | |
Liquid | |
(metals basis) |
Colorless to Yellow | |
∼102°C | |
1.384 | |
MFCD00008280 | |
UN1835 | |
Air Sensitive | |
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide | |
WGTYBPLFGIVFAS-UHFFFAOYSA-M | |
tetramethylazanium;hydroxide | |
60966 | |
99.9999% | |
25% w/w aq. soln. | |
Tetramethylammonium hydroxide |
Safety and Handling
GHS H Statement
H300-H310-H370-H372-H314
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301-H310-H314-H370-H372
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: TETRAMETHYLAMMONIUM HYDROXIDE SOLUTION
EINECSNumber : 200-882-9
RTECSNumber : PA0875000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only