missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium hydroxide, 25 wt% in methanol
CAS: 75-59-2 | C4H13NO | 91.154 g/mol
$119.94 - $304.50
Specifications
| Chemical Name or Material | Tetramethylammonium hydroxide |
|---|---|
| Name Note | 25 wt. % in Methanol |
| CAS | 75-59-2,67-56-1 |
| Boiling Point | 65.0°C |
| Melting Point | -98.0°C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC138351000
|
Thermo Scientific Chemicals
138351000 |
100 g | Plastic bottle |
Each for $119.94
|
|
||||
|
AC138355000
|
Thermo Scientific Chemicals
138355000 |
500 g | Plastic bottle |
Each for $304.50
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
| Tetramethylammonium hydroxide | |
| 75-59-2,67-56-1 | |
| -98.0°C | |
| (CH3)4NOH | |
| 0.9000g/mL | |
| Solubility in water: soluble. | |
| C[N+](C)(C)C.[OH-] | |
| 91.154 | |
| 91.15 | |
| 6°C |
| 25 wt. % in Methanol | |
| 65.0°C | |
| C4H13NO | |
| MFCD00008280 | |
| tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide | |
| WGTYBPLFGIVFAS-UHFFFAOYSA-M | |
| tetramethylazanium;hydroxide | |
| 60966 | |
| 0.9 |
RUO – Research Use Only