Learn More
Tetramethylammonium hydroxide pentahydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 207520250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetramethylammonium hydroxide pentahydrate | |
62.0°C to 71.0°C | |
Authentic | |
Glass bottle | |
(CH3)4NOH·5H2O | |
25 g | |
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm | |
MYXKPFMQWULLOH-UHFFFAOYSA-M | |
tetramethylazanium;hydroxide;pentahydrate | |
82620 | |
97% |
10424-65-4 | |
White | |
97% | |
C4H23NO6 | |
MFCD00149566 | |
04, 50 | |
Solubility in water: soluble. | |
O.O.O.O.O.[OH-].C[N+](C)(C)C | |
181.23 | |
181.23 | |
Hygroscopic Crystals |
Chemical Identifiers
10424-65-4 | |
181.23 | |
MYXKPFMQWULLOH-UHFFFAOYSA-M | |
82620 | |
O.O.O.O.O.[OH-].C[N+](C)(C)C |
C4H23NO6 | |
MFCD00149566 | |
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm | |
tetramethylazanium;hydroxide;pentahydrate |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Toxic to aquatic life w
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Danger
RUO – Research Use Only