missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylammonium hydroxide pentahydrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 420531000

 View more versions of this product

Catalog No. AC420531000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetramethylammonium hydroxide pentahydrate
68°C to 71°C
Authentic
Glass Bottle
(CH3)4NOH·5H2O
100 g
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
O.O.O.O.O.[OH-].C[N+](C)(C)C
181.23
181.23
Crystalline Powder
10424-65-4
White
99%
C4H23NO6
MFCD00149566
04, 50
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylazanium;hydroxide;pentahydrate
82620
≥98.5%

Chemical Identifiers

10424-65-4
181.23
MYXKPFMQWULLOH-UHFFFAOYSA-M
82620
O.O.O.O.O.[OH-].C[N+](C)(C)C
C4H23NO6
MFCD00149566
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
tetramethylazanium;hydroxide;pentahydrate
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes severe skin burns and eye damage. Toxic if swallowed. Toxic in contact with skin.

GHS P Statement Do not breathe dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact le

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only